MilliporeSigma
Buffer Reference Center
Choosing the Right Biological Buffer
Choose a buffer based on your pH requirements as well as the pKa, a measure of acid strength that accounts for pH, concentration, and temperature. Regulatory or purity needs for your exact application should also be considered. The following tables can help you navigate preparation of many common buffer solutions by pH and pKa. 
In addition to the tables below, we've developed several buffer recipe calculators to assist your buffer preparation.
TBE and TAE for gel electrophoresis have slightly more involved recipes. We can help you pick the right one.
What are buffer grades?
Buffer grade indicates the quality and impurity levels appropriate for different uses. We provide six grades of buffers indicated for general lab use, final pharmaceutical formulation and manufacturing, and applications in between that may need trace metal testing or materials of a specified purity.
Useful pH Ranges of Selected Biological Buffers Chart (25 °C, 0.1 M)  
BuffersUseful pH RangepKa
(at 20 °C)
pKa
(at 25 °C)
pKa
(at 37 °C)
MES5.5–6.76.166.105.97
Bis-Tris5.8–7.2n/a6.506.36
ADA6.0–7.26.656.596.46
ACES6.1–7.56.886.786.54
PIPES6.1–7.56.806.766.66
MOPSO6.2–7.6n/a6.906.75
Bis-Tris Propane6.3–9.5n/a6.8, 9.0n/a
BES6.4–7.87.177.096.90
MOPS6.5–7.97.287.207.02
TES6.8–8.27.507.407.16
HEPES6.8–8.27.557.487.31
DIPSO7.0–8.2n/a7.607.35
MOBS6.9–8.3n/a7.60n/a
TAPSO7.0–8.2n/a7.607.39
Tris or Trizma®7.0–9.08.208.067.72
HEPPSO7.1–8.5n/a7.806.66
POPSO7.2–8.5n/a7.807.63
TEA7.3–8.3n/a7.80n/a
EPPS7.3–8.7n/a8.00n/a
Tricine7.4–8.88.168.057.80
Gly-Gly7.5–8.9n/a8.20n/a
Bicine7.6–9.08.358.268.04
HEPBS7.6–9.0n/a8.30n/a
TAPS7.7-9.18.498.408.18
AMPD7.8–9.7n/a8.80n/a
TABS8.2–9.6n/a8.90n/a
AMPSO8.3–9.7n/a9.009.10
CHES8.6–10.09.559.499.36
CAPSO8.9–10.3n/a9.609.43
AMP9.0–10.5n/a9.70n/a
CAPS9.7–11.110.5610.4010.02
CABS10.0–11.4n/a10.70n/a
Tris or Trizma® Buffer Preparation Table – pH vs. Temperature
pH at Temperature
5 °C25 °C37 °C
7.767.206.91
7.897.307.02
7.977.407.12
8.077.507.22
8.187.607.30
8.267.707.40
8.377.807.52
8.487.907.62
8.588.007.71
8.688.107.80
8.788.207.91
8.888.308.01
8.988.408.10
9.098.508.22
9.188.608.31
9.288.708.42
9.368.808.51
9.478.908.62
9.569.008.70
g/L for 0.05 M Solution
Trizma® HCLTrizma® Base
7.020.67
6.850.80
6.610.97
6.351.18
6.061.39
5.721.66
5.321.97
4.882.30
4.442.65
4.022.97
3.543.34
3.073.70
2.644.03
2.214.36
1.834.65
1.504.90
1.235.13
0.965.32
0.765.47
Phosphate Buffer Preparation Table – 0.2 M solution  
Potassium 
Phosphate Monobasic Anhydrous g/L
Sodium Phosphate Dibasic Heptahydrate g/L23 °C pH
22.43.495.7
22.084.295.8
21.605.375.9
21.056.606.0
20.408.056.1
19.569.936.2
18.6012.076.3
17.6414.226.4
16.4416.906.5
15.0020.126.6
13.5623.346.7
12.2426.296.8
Potassium Phosphate Monobasic
Anhydrous g/L
Sodium Phosphate
Dibasic Heptahydrate g/L
23 °C pH
10.8029.516.9
9.3632.737.0
7.9235.957.1
6.7238.637.2
5.5241.317.3
4.5643.467.4
3.8445.077.5
3.1246.687.6
2.5248.557.7
2.0449.097.8
1.6849.897.9
1.2750.818.0
Citric Acid – Na2HPO4 Buffer Preparation, pH 2.6–7.61
Citric acid monohydrate, C6H8O7 • H2O, MW 210.14; 0.1 M contains 21.01 g/L. Na2HPO4, MW 141.98; 0.2 M contains 28.40 g/L, or Na2HPO4 • 2H2O, MW 178.05; 0.2 M contains 35.61 g/L.
x mL 0.1 M-citric acid and y mL 0.2 M-Na2HPO4 mixed
pHx mL 0.1 M-citric acidy mL 0.2-Na2HPO4
2.689.1010.90
2.884.1515.85
3.079.4520.55
3.275.3024.70
3.471.5028.50
3.667.8032.20
3.864.5035.50
4.061.4538.55
4.258.6041.40
4.455.9044.10
4.653.2546.75
4.850.7049.30
5.048.5051.50
5.246.4053.60
5.444.2555.75
5.642.0058.00
5.839.5560.45
6.036.8563.15
6.233.9066.10
6.430.7569.25
6.627.2572.75
6.822.7577.25
7.017.6582.35
7.213.0586.95
7.49.1590.85
7.66.3593.65
Citric Acid – Sodium Citrate Buffer Preparation, pH 3.0-6.21
Citric acid monohydrate, C6H8O7 • H2O, MW 210.14; 0.1 M contains 21.01 g/L. Trisodium citrate dihydrate, C6H5O7Na3 • 2H2O, MW 294.12; 0.1 M contains 29.41 g/L.
x mL 0.1 M-citric acid and y mL 0.1 M-trisodium citrate mixed
pHx mL 0.1 M-citric acidy mL 0.1 M-trisodium citrate
3.082.018.0
3.277.522.5
3.473.027.0
3.668.531.5
3.863.536.5
4.059.041.0
4.254.046.0
4.449.550.5
4.644.555.5
4.840.060.0
5.035.065.0
5.230.569.5
5.425.574.5
5.621.079.0
5.816.084.0
6.011.588.5
6.28.092.0
Sodium Acetate – Acetic Acid Buffer PREPARATION, pH 3.7–5.61
Sodium acetate trihydrate, CH3COONa • 3H2O, MW 136.09; 0.2 M contains 27.22 g/L.
x mL 0.2 M-NaOAc and y mL 0.2 M-HOAc mixed
pH, 18 °Cx mL 0.2 M-NaOAcy mL 0.2 M-HOAc
3.710.090.0
3.812.088.0
4.018.082.0
4.226.573.5
4.437.063.0
4.649.051.0
4.859.041.0
5.070.030.0
5.279.021.0
5.486.014.0
5.691.09.0
Na2HPO4 – NaH2PO4 Buffer PREPARATION, pH 5.8-8.0 at 25 °C1
Na2HPO4 • 2H2O, MW 178.05; 0.2 M contains 35.61 g/L. Na2HPO4 • 12H2O, MW 358.22; 0.2 M contains 71.64 g/L. NaH2PO4 • H2O, MW 138.01; 0.2 M contains 27.6 g/L. NaH2PO4 • 2H2O, MW 156.03; 0.2 M contains 31.21 g/L.
x mL 0.2 M-Na2HPO4, y mL 0.2 M-NaH2PO4; diluted to 100 mL with H2O
pH, 25 °Cx mL 0.2 M-Na2HPO4y mL 0.2 M-NaH2PO4
5.84.046.0
6.06.1543.85
6.29.2540.75
6.413.2536.75
6.618.7531.25
6.824.525.5
7.030.519.5
7.236.014.0
7.440.59.5
7.643.56.5
7.845.754.25
8.047.352.65
Imidazole (glyoxaline) – HCl Buffer PREPARATION, pH 6.2–7.8 at 25 °C1
Imidazole, C3H4N2, MW 68.08
25 mL 0.2 M-imidazole (13.62 g/L), x mL 0.2 M-HCl, diluted to 100 mL with H2O
pH, 25 °Cx mL 0.2 M-HCl
6.221.45
6.419.9
6.617.75
6.815.2
7.012.15
7.29.3
7.46.8
7.64.65
7.83.0
Sodium Carbonate – Sodium Bicarbonate Buffer PREPARATION, pH 9.2-10.81
Na2CO3 • 10H2O, MW 286.2; 0.1 M contains 28.62 g/L. NaHCO3, MW 84.0; 0.1 M contains 8.40 g/L.
x mL 0.1 M-Na2CO3 and y mL 0.1 M-Na2HCO3 mixed
20 °C37 °C  
x mL 0.1 M-Na2CO3y mL 0.1 M-Na2HCO3  
9.28.81090
9.49.12080
9.59.43070
9.89.54060
9.99.75050
10.19.96040
10.310.17030
10.510.38020
10.810.69010
Buffer Preparation Formulas and Equations
Percentage by weight (w/v)
(% buffer desired / 100) × final buffer volume (mL) = g of starting material needed.
Molar Solutions
desired molarity × formula weight × solution final volume (L) = grams needed
Henderson-Hasselbach Equation
Henderson ph
Reference
1.
Dawson R, Elliot D, Elliot W, Jones KM. 1986. Data for Biochemical Research. 3rd ed.. Oxford Science Publ..